CAS RN 910232-84-7
IUPAC Name:
4-tert-butyl-N-[2-methyl-3-(4-methyl-6-{[4-(morpholine-4-carbonyl)phenyl]amino}-5-oxo-4,5-dihydropyrazin-2-yl)phenyl]benzamide
Synonyms:
4-tert-butyl-N-[2-methyl-3-(4-methyl-6-{[4-(morpholine-4-carbonyl)phenyl]amino}-5-oxopyrazin-2-yl)phenyl]benzamide
Molecular Weight (g/mol):
579.7
Molecular Formula:
C34H37N5O4
InChi Key:
JIFCFQDXHMUPGP-UHFFFAOYSA-N
SMILES:
CN1C=C(N=C(NC2=CC=C(C=C2)C(=O)N2CCOCC2)C1=O)C1=C(C)C(NC(=O)C2=CC=C(C=C2)C(C)(C)C)=CC=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals CGI1746, 95%
CAS: 910232-84-7 Molecular Formula: C34H37N5O4
2
R&D Systems™ CGI 1746
A Tocris product, part of R&D Systems. Potent, reversible BTK inhibitor