CAS RN 911222-45-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ LY 2603618
Potent and selective Chk1 inhibitor (IC50 = 7 nM)