accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91271-59-9

IUPAC Name: {3-[(pyrrolidin-1-yl)methyl]phenyl}methanol
Synonyms: [3-(pyrrolidin-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: YMFREPVLJDRBOG-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(CN2CCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results