accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOH

CAS RN 91271-62-4

IUPAC Name: {4-[(piperidin-1-yl)methyl]phenyl}methanol
Synonyms: [4-(piperidin-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: SQWKPHFBCVNPGU-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results