accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCH3OHH3CCH3

CAS RN 913-86-0

IUPAC Name: tetrakis(4-methylphenyl)ethane-1,2-diol
Synonyms: tetrakis(4-methylphenyl)ethane-1,2-diol
Molecular Weight (g/mol): 422.57
Molecular Formula: C30H30O2
InChi Key: UWVQXJFNNKWNLZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(O)(C1=CC=C(C)C=C1)C(O)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results