accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNHFOS

CAS RN 913186-74-0

IUPAC Name: N-(4-fluorophenyl)-4-methyl-5-(thiophene-3-carbonyl)-1,3-thiazol-2-amine
Synonyms: N-(4-fluorophenyl)-4-methyl-5-(thiophene-3-carbonyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 318.38
Molecular Formula: C15H11FN2OS2
InChi Key: PEAMDZVDNYENPN-UHFFFAOYSA-N
SMILES: CC1=C(SC(NC2=CC=C(F)C=C2)=N1)C(=O)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)