CAS RN 91374-20-8
IUPAC Name:
hydrogen 4-[2-(dipropylamino)ethyl]-2,3-dihydro-1H-indol-2-one chloride
Synonyms:
ropinirole
Molecular Weight (g/mol):
296.84
Molecular Formula:
C16H25ClN2O
InChi Key:
XDXHAEQXIBQUEZ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CCCN(CCC)CCC1=CC=CC2=C1CC(=O)N2
1
–
3
of
3
results
1
Promotions Available
Thermo Scientific Chemicals Ropinirole hydrochloride
Thermo Scientific Chemicals Ropinirole hydrochloride
CAS: 91374-20-8 Molecular Formula: C16H24N2O·HCl
2
Ropinirole Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
3
R&D Systems™ Ropinirole
CAS: 91374-20-8 Molecular Formula: C16H25ClN2O