accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-69-5

IUPAC Name: [5-(dihydroxyboranyl)-1-benzofuran-2-yl]boronic acid
Synonyms: 5-(dihydroxyboranyl)-1-benzofuran-2-ylboronic acid
Molecular Weight (g/mol): 205.77
Molecular Formula: C8H8B2O5
InChi Key: LCHGNXMXVDSVAR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC(=CC=C2O1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results