accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOClO

CAS RN 91454-65-8

IUPAC Name: 2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]acetyl chloride
Synonyms: 2-[(4-methyl-2-oxochromen-7-yl)oxy]acetyl chloride
Molecular Weight (g/mol): 252.65
Molecular Formula: C12H9ClO4
InChi Key: CTSWCAAFHPVFFO-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(OCC(Cl)=O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results