accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91510-62-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 0
Molecular Formula: —
InChi Key: —
SMILES: *
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)