Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Galacturonic Acid Monohydrate 99%, Thermo Scientific™

Catalog No. AC227820250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227820250 25 g
AC227820050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227820250 Supplier Thermo Scientific Chemicals Supplier No. 227820250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91510-62-2
Molecular Formula C6H9O7
Molecular Weight (g/mol) 0.00
InChI Key AEMOLEFTQBMNLQ-DTEWXJGMSA-M
Synonym d-+-galacturonic acid monohydrate
PubChem CID 91872458
IUPAC Name (2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES *

Specifications

Color White to Beige
Infrared Spectrum Authentic
Quantity 25 g
Specific Rotation After 1 hr., on dried substance: +51° to +55° (20°C, 589nm) (c=4, H2O)
Formula Weight 212.16
Percent Purity ≥98.5%
Physical Form Fine Powder
Chemical Name or Material D(+)-Galacturonic acid monohydrate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.