accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNH2

CAS RN 915707-48-1

IUPAC Name: 1-{3-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl}methanamine
Synonyms: 1-{3-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl}methanamine
Molecular Weight (g/mol): 233.36
Molecular Formula: C14H23N3
InChi Key: MSLZRDIUXASDJH-UHFFFAOYSA-N
SMILES: CN1CCCN(CC2=CC(CN)=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results