accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNSONONH

CAS RN 916141-36-1

IUPAC Name: N-{[1,1'-biphenyl]-2-yl}-1-{2-[(1-methyl-1H-1,3-benzodiazol-2-yl)sulfanyl]acetyl}pyrrolidine-2-carboxamide
Synonyms: N-{[1,1'-biphenyl]-2-yl}-1-{2-[(1-methyl-1,3-benzodiazol-2-yl)sulfanyl]acetyl}pyrrolidine-2-carboxamide
Molecular Weight (g/mol): 470.59
Molecular Formula: C27H26N4O2S
InChi Key: YSBGRVXJEMSEQY-UHFFFAOYNA-N
SMILES: CN1C(SCC(=O)N2CCCC2C(=O)NC2=CC=CC=C2C2=CC=CC=C2)=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)