accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 916766-97-7

IUPAC Name: 2-(thiophen-2-yl)pyrimidine-5-carboxylic acid
Synonyms: 2-(thiophen-2-yl)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 206.22
Molecular Formula: C9H6N2O2S
InChi Key: SJJLOOUOVNRKLF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results