accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 917-54-4

IUPAC Name: methyllithium
Synonyms: methyllithium|LiMe
Molecular Weight (g/mol): 21.98
Molecular Formula: CH3Li
InChi Key: DVSDBMFJEQPWNO-UHFFFAOYSA-N
SMILES: [Li]C
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results