accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91898-93-0

IUPAC Name: 2-(4-{bromobis[4-(5,6-dichloro-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)phenyl]methyl}phenyl)-5,6-dichloro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(4-{bromobis[4-(5,6-dichloro-1,3-dioxoisoindol-2-yl)phenyl]methyl}phenyl)-5,6-dichloroisoindole-1,3-dione
Molecular Weight (g/mol): 965.24
Molecular Formula: C43H18BrCl6N3O6
InChi Key: JYHNGHXQORJDGQ-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C2C(=O)N(C(=O)C2=C1)C1=CC=C(C=C1)C(Br)(C1=CC=C(C=C1)N1C(=O)C2=CC(Cl)=C(Cl)C=C2C1=O)C1=CC=C(C=C1)N1C(=O)C2=CC(Cl)=C(Cl)C=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results