accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CONCH3NOCH3OH3COOH3C

CAS RN 91940-87-3

IUPAC Name: hydrogen 3-(3-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}propyl)-7,8-dimethoxy-2,3,4,5-tetrahydro-1H-3-benzazepin-2-one chloride
Synonyms: hydrogen 3-(3-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}propyl)-7,8-dimethoxy-4,5-dihydro-1H-3-benzazepin-2-one chloride
Molecular Weight (g/mol): 493.04
Molecular Formula: C26H37ClN2O5
InChi Key: ZRNKXJHEQKMWCH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(CCN(C)CCCN2CCC3=CC(OC)=C(OC)C=C3CC2=O)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results