accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-78-4

IUPAC Name: N-(4-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
Synonyms: N-(4-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
Molecular Weight (g/mol): 297.74
Molecular Formula: C17H12ClNO2
InChi Key: OHAXNCGNVGGWSO-UHFFFAOYSA-N
SMILES: OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=C(Cl)C=C1
Form 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results