Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-3-hydroxy-2-naphthanilide 97.0+%, TCI America™
SDP

Catalog No. C210325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C210325G Supplier TCI America Supplier No. C210325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-78-4
Molecular Formula C17H12ClNO2
Molecular Weight (g/mol) 297.738
MDL Number MFCD00021639
InChI Key OHAXNCGNVGGWSO-UHFFFAOYSA-N
Synonym N-(4-Chlorophenyl)-3-hydroxy-2-naphthamide, Naphthol AS-E, Azoic Coupling Component 10
PubChem CID 66720
IUPAC Name N-(4-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)NC3=CC=C(C=C3)Cl)O

Specifications

Melting Point 264°C
Quantity 25 g
Formula Weight 297.74
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-3-hydroxy-2-naphthanilide
EINECSNumber (5)-2279
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.