accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2

CAS RN 92-87-5

IUPAC Name: [1,1'-biphenyl]-4,4'-diamine
Synonyms: benzidine 4,4'-diamonophenyl benzidine (substance) benzidina 4,4'-diaminodiphenyl p,p-bianiline benzioine C.I. azoic diazo component 112 benzidine base benzydyna
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: HFACYLZERDEVSX-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results