Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzidine, Restek, 99%, in Methanol
SDP

Catalog No. 06801698 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-801-698 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-801-698 Supplier Restek Corporation Supplier No. 31441
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Benzidine 1000μg/mL, Methanol, 1mL/ampule

Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements

Chemical Identifiers

CAS 92-87-5 , 67-56-1
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
InChI Key HFACYLZERDEVSX-UHFFFAOYSA-N

Specifications

Physical Form Liquid
Type Certified Reference Standard for Chromatography
Quantity 1 mL
Percent Purity 99%
Chemical Name or Material Certified Reference Standard
ShelfLife Max Shelf Life on Ship Date: 55 months; Min Shelf Life on Ship Date: 6 months
Recommended Storage 10°C or colder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.