Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzidine Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USEPA1071
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1071 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1071 Supplier Agilent Technologies Supplier No. EPA10711
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-87-5 , 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym benzidine, 4,4'-diaminobiphenyl, p-diaminodiphenyl, 4,4'-bianiline, 1,1'-biphenyl-4,4'-diamine, 4,4'-biphenyldiamine, 4,4'-diaminodiphenyl, biphenyl-4,4'-diamine, 4,4'-biphenylenediamine, 4,4'-diphenylenediamine
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 0.63, 99.36
Analyte or Component Names methanol,aldrin
Element Methanol,aldrin
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Benzidine Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.