accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 920-46-7

IUPAC Name: 2-methylprop-2-enoyl chloride
Synonyms: methacryloyl chloride
Molecular Weight (g/mol): 104.53
Molecular Formula: C4H5ClO
InChi Key: VHRYZQNGTZXDNX-UHFFFAOYSA-N
SMILES: CC(=C)C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)