accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONClOH

CAS RN 92172-83-3

IUPAC Name: (2-chloro-6-methoxyquinolin-3-yl)methanol
Synonyms: (2-chloro-6-methoxyquinolin-3-yl)methanol
Molecular Weight (g/mol): 223.66
Molecular Formula: C11H10ClNO2
InChi Key: AKKPSGFLKITYGV-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(Cl)C(CO)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results