Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxyquinoline-3-methanol, 97%, Thermo Scientific™

Catalog No. MO07164DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07164DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07164DA Supplier Thermo Scientific Chemicals Supplier No. MO07164DA

Chemical Identifiers

CAS 92172-83-3
Molecular Formula C11H10ClNO2
Molecular Weight (g/mol) 223.656
MDL Number MFCD02232332
InChI Key AKKPSGFLKITYGV-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxyquinoline-3-methanol, 2-chloro-6-methoxyquinolin-3-yl methanol, 2-chloro-6-methoxy-quinolin-3-yl-methanol, 2-chloro-6-methoxy-3-quinolyl methan-1-ol, 3-quinolinemethanol,2-chloro-6-methoxy, 3-quinolinemethanol, 2-chloro-6-methoxy, 2-chloro-6-methoxy-3-quinolinyl methanol, 2-chloro-3-hydroxymethyl-6-methoxyquinoline, 2-chloranyl-6-methoxy-quinolin-3-yl methanol
PubChem CID 865110
IUPAC Name (2-chloro-6-methoxyquinolin-3-yl)methanol
SMILES COC1=CC2=CC(=C(N=C2C=C1)Cl)CO

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 223.66
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methoxyquinoline-3-methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.