accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)S(R)NH3OOOO

CAS RN 922-55-4

IUPAC Name: (2S)-2-azaniumyl-3-{[(2R)-2-azaniumyl-2-carboxylatoethyl]sulfanyl}propanoate
Synonyms: (2S)-2-ammonio-3-{[(2R)-2-ammonio-2-carboxylatoethyl]sulfanyl}propanoate
Molecular Weight (g/mol): 208.23
Molecular Formula: C6H12N2O4S
InChi Key: DWPCPZJAHOETAG-ZXZARUISSA-N
SMILES: [NH3+][C@@H](CSC[C@@H]([NH3+])C([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)