CAS RN 923564-51-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ABT 263
High affinity Bcl-2 family inhibitor; proapoptotic, senolytic and antitumor