accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 924-88-9

IUPAC Name: 1,4-bis(propan-2-yl) butanedioate
Synonyms: 1,4-diisopropyl butanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: YPLYFEUBZLLLIY-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CCC(=O)OC(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results