accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH

CAS RN 925-94-0

IUPAC Name: formic acid
Synonyms: formic acid ameisensaeure hcooh acide formique bilorin H-cooh
Molecular Weight (g/mol): 46.03
Molecular Formula: CH2O2
InChi Key: BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES: OC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results