CAS RN 925393-51-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[3-(Trifluoromethyl)phenyl]acetamidoxime, 97%, Thermo Scientific™
CAS: 925393-51-7