accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926-77-2

IUPAC Name: 2-(2-aminoacetamido)propanoic acid
Synonyms: 2-(2-aminoacetamido)propanoic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: VPZXBVLAVMBEQI-UHFFFAOYNA-N
SMILES: CC(NC(=O)CN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results