CAS RN 926908-04-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Histone Deacetylase Inhibitor VI, HNHA
The Histone Deacetylase Inhibitor VI, HNHA, also referenced under CAS 926908-04-5, controls the biological activity of Histone Deacetylase.