accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926921-67-7

IUPAC Name: 1-{4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Synonyms: 1-{4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Molecular Weight (g/mol): 215.26
Molecular Formula: C12H13N3O
InChi Key: JNWHMYUBZLJKJT-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(CN)C=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results