accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOONHSSHNSOOClClCl

CAS RN 927822-86-4

IUPAC Name: 2,5-dichloro-N-(2-{[2-(2,5-dichlorobenzenesulfonamido)ethyl]disulfanyl}ethyl)benzene-1-sulfonamide
Synonyms: 2,5-dichloro-N-(2-{[2-(2,5-dichlorobenzenesulfonamido)ethyl]disulfanyl}ethyl)benzenesulfonamide
Molecular Weight (g/mol): 570.35
Molecular Formula: C16H16Cl4N2O4S4
InChi Key: REQLACDIZMLXIC-UHFFFAOYSA-N
SMILES: ClC1=CC(=C(Cl)C=C1)S(=O)(=O)NCCSSCCNS(=O)(=O)C1=C(Cl)C=CC(Cl)=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results