CAS RN 927827-98-3
CAS RN 927827-98-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MMP Inhibitor III
The MMP Inhibitor III, also referenced under CAS 927827-98-3, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.