accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(S)H2N

CAS RN 92932-74-6

IUPAC Name: (1S)-1,2,3,4-tetrahydroisoquinolin-2-ium-1-carboxylate
Synonyms: (1S)-1,2,3,4-tetrahydroisoquinolin-2-ium-1-carboxylate
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: OXFGRWIKQDSSLY-VIFPVBQESA-N
SMILES: [O-]C(=O)[C@H]1[NH2+]CCC2=CC=CC=C12
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results