Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Catalog No. AC326132500
Change view
Click to view available options
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC326132500 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC326132500 Supplier Thermo Scientific Chemicals Supplier No. 326132500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 92932-74-6
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H12ClNO2
Quantity 250mg
Synonym 1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid hydrochloride, dl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid hydrochloride, 1,2,3,4-tetrahydroisoquinolinecarboxylic acid, chloride, 1-tic.hcl, d,l-1-tic hcl, akos 46, timtec-bb sbb003410, cambridge id 5926716, 1s-3,4-dihydro-1h-isoquinoline-1-carboxylate, dl-1,2,3,4-tetrahydroisoquinoline-1-carboxylicacidhcl
InChI Key PMPYPSNDPNMYIO-UHFFFAOYSA-N
SMILES C1CNC(C2=CC=CC=C21)C(=O)O.Cl
IUPAC Name 1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 213.661
PubChem CID 2733448
Formula Weight 213.66
Percent Purity ≥96.0% (on dry substance)
Chemical Name or Material DL-1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.