accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 931-56-6

IUPAC Name: methoxycyclohexane
Synonyms: cyclohexane, methoxy-
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: GHDIHPNJQVDFBL-UHFFFAOYSA-N
SMILES: COC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results