Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxycyclohexane 98.0+%, TCI America™
SDP

Catalog No. M23745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23745G 5 g
M237425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23745G Supplier TCI America Supplier No. M23745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 931-56-6
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00154087
InChI Key GHDIHPNJQVDFBL-UHFFFAOYSA-N
PubChem CID 13607
IUPAC Name methoxycyclohexane
SMILES COC1CCCCC1

Specifications

Melting Point -74°C
Color Colorless
Boiling Point 135°C
Quantity 5 g
UN Number 3271
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxycyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.