accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3PNCH3H3CCH3CH3H3C

CAS RN 932710-63-9

IUPAC Name: 4-(di-tert-butylphosphanyl)-N,N-dimethylaniline
Synonyms: 4-(di-tert-butylphosphanyl)-N,N-dimethylaniline
Molecular Weight (g/mol): 265.38
Molecular Formula: C16H28NP
InChi Key: IQTHEAQKKVAXGV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)