CAS RN 934389-88-5
IUPAC Name:
2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one hydrochloride
Synonyms:
2-(morpholin-4-yl)-8-phenylchromen-4-one hydrochloride
Molecular Weight (g/mol):
343.81
Molecular Formula:
C19H18ClNO3
InChi Key:
OQZQSRICUOWBLW-UHFFFAOYSA-N
SMILES:
Cl.O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1
1
–
1
of
1
results
1
Tocris Bioscience™ LY 294002 hydrochloride
CAS: 934389-88-5 Molecular Formula: C19H18ClNO3