accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93457-07-9

CH3OClCl

CAS RN 93457-07-9

IUPAC Name: 1-(2,4-dichlorophenyl)propan-2-one
Synonyms: 1-(2,4-dichlorophenyl)propan-2-one
Molecular Weight (g/mol): 203.06
Molecular Formula: C9H8Cl2O
InChi Key: ITVVHXZQWSCZBC-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results