CAS RN 935666-88-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ AZD 1480
Potent and selective JAK2 inhibitor; antiangiogenic