accessibility menu, dialog, popup

UserName

Viewing profile as user:
POBrBr

CAS RN 93869-52-4

IUPAC Name: 1-bromo-4-[(4-bromophenyl)(phenyl)phosphoroso]benzene
Synonyms: 1-bromo-4-[4-bromophenyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 436.08
Molecular Formula: C18H13Br2OP
InChi Key: URGNHDJCYWEAKG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)P(=O)(C1=CC=CC=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results