CAS RN 939825-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ OB 24 hydrochloride
Potent and selective heme oxygenase 1 (HO-1) inhibitor