accessibility menu, dialog, popup

UserName

Viewing profile as user:
OON

CAS RN 94-44-0

IUPAC Name: benzyl pyridine-3-carboxylate
Synonyms: benzyl nicotinate rubriment niacin benzyl ester nicotinsaeurebenzylester pykaryl benzyl nicotinate - chemical pycaril benzylis nicotinas estru benzylowego kwasu nikotynowego
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: KVYGGMBOZFWZBQ-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C1=CC=CN=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (7)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (5)
  • (2)
Physical Form 
  • (1)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (5)