Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Nicotinate, Approx. 99%, Spectrum™ Chemical
SDP

Catalog No. 18609063 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-063 50 g Amber Glass Bottle
18-609-061 250 g Amber Glass Bottle
18-609-059 1 kg Amber Glass Bottle
18-609-060 2.5 kg Amber Glass Bottle
18-609-062 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-063 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B121250GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-44-0

Specifications

CAS Max % 1
Boiling Point 189°C (372°F) at 1.6 kPa
Packaging Amber Glass Bottle
Quantity 50 g
Molecular Weight (g/mol) 213.23
Percent Purity ∼99%
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Benzyl Nicotinate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.