Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Nicotinate, MP Biomedicals™

Catalog No. MP520933980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520933980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP520933980 Supplier MP Biomedicals, Inc Supplier No. 0520933980

missing translation for 'validMainframeMsg'

Benzyl Nicotinate ≥98%, Yellowish Oily Liquid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-44-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00023584
InChI Key KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Synonym Nicotinic acid benzyl ester

Specifications

Melting Point 24°C (literature)
Color Yellow
Boiling Point 177°C (8 mmHg) (Literature)
Flash Point 113°C / 235.4°F (lit.)
Refractive Index n20/D 1.570 (lit.)
Quantity 100 g
Beilstein 159169
Physical Form Oily Liquid
Grade ≥98%
RTECSNumber QT0850000
storageNote1 Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.