accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOOHO

CAS RN 94-81-5

IUPAC Name: 4-(4-chloro-2-methylphenoxy)butanoic acid
Synonyms: MCPB
Molecular Weight (g/mol): 228.67
Molecular Formula: C11H13ClO3
InChi Key: LLWADFLAOKUBDR-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1OCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results