accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3H3C

CAS RN 94-98-4

IUPAC Name: (2,4-dimethylphenyl)methanaminium
Synonyms: (2,4-dimethylphenyl)methanaminium
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: GBSUVYGVEQDZPG-UHFFFAOYSA-O
SMILES: CC1=CC=C(C[NH3+])C(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)